Order Entry
Denmark
ContactUsLinkComponent
Anti-HFM1 Rabbit Polyclonal Antibody (HRP)
Anti-HFM1 Rabbit Polyclonal Antibody (HRP)
  BOSSBS-15471R-HRP
 :  Bioss
Anti-HFM1 Rabbit Polyclonal Antibody (HRP)
  BOSSBS-15471R-HRP
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    HFM1
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    151-250/1435
  • Form:
    Liquid
  • Antigen synonyms:
    HFM1|SEC63 domain-containing protein 1|HFM1_HUMAN|Probable ATP-dependent DNA helicase HFM1
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    163 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human HFM1
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

HFM1.

Type: Primary
Antigen: HFM1
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 151-250/1435
Host: Rabbit
Isotype: IgG
Reactivity: