Anti-C6ORF151 Rabbit Polyclonal Antibody (HRP)
Catalog # BOSSBS-15225R-HRP
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:C6orf151
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Epitope:151-250/339
- Form:Liquid
- Antigen synonyms:U11/U12 small nuclear ribonucleoprotein 48 kDa protein|Snrnp48|U11/U12 snRNP 48 kDa protein|SNR48_HUMAN|U11/U12-48K|Small nuclear ribonucleoprotein 48 kDa
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:40 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human C6ORF151
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
C6orf151.
Type: Primary
Antigen: C6orf151
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 151-250/339
Host: Rabbit
Isotype: IgG
Reactivity: