Specifications
- Antibody type:Primary
- Antigen name:Dynamin 1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:551-680/864
- Form:Liquid
- Gene ID:1759
- Antigen synonyms:DYN1_HUMAN|DNM1|Dynamin-1|B dynamin|DNM|DNM 1|Dynamin|Dynamin1|D100
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:97 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Dynamin 1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Dynamin I is a GTPase enzyme required for the retrieval of synaptic vesicles after exocytosis and functions in endocytosis by stimulating assembly of invaginating synaptic vesicles (1). Dynamin I is phosphorylated in nerve terminals exclusively in the cytosolic compartment and in vitro by protein kinase C (PKC) (25). The phosphorylation site in PKC-phosphorylated Dynamin I is a single site at Serine 795, which is located near a binding site for the SH3 domain of p85, the regulatory subunit of phosphatidylinositol 3-kinase (25). Dephosphorylation is required for synaptic vesicle retrieval, suggesting that phosphorylation affects the subcellular localization of Dynamin I (5). Mouse, rat and human Dynamin I are phosphorylated on serine residues, including Ser 778, by Cdk5, regulating PACSIN1 recruitment and enabling synaptic vesicle endocytosis.
Type: Primary
Antigen: Dynamin 1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 551-680/864
Host: Rabbit
Isotype: IgG
Reactivity: