Anti-CDK4 Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-31308
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Cyclin-dependent Kinase 4
- Antigen symbol:CDK4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Bovine,Rat,Mouse
- Western blot:Yes
- Gene ID:1019
- Antigen synonyms:CMM3|MGC14458|cyclin-dependent kinase 4|Cell division protein kinase 4|EC 2.7.11.22|PSK-J3cell division kinase 4|EC 2.7.11|3|melanoma cutaneous malignant
- Storage buffer:PBS, 20% Glycerol (pH7),
- Storage temperature:Aliquot and store at ‒20 °C or ‒80 °C. Avoid freeze-thaw cycles.
- Concentration:1,3 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human CDK4. The exact sequence is proprietary.
- Purification:Immunogen affinity purified
- Pk:100 µl
Specifications
About this item
The CDK4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CDK4. This antibody reacts with human, mouse, rat, bovine. The CDK4 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100 to 1:500
Type: Primary
Antigen: CDK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine