Ascorbate Assay Kit
Catalog # CAYM700420-96
Supplier: Cayman Chemical
CAS Number:
Specifications
- Pk:1
- Description:Kit contains: Standard, buffer, DTPA, substrate, developer, and pate, range 5 - 150 µM
- Brand name:CAYM
Specifications
About this item
A kit for measuring ascorbate.
- Quantify ascorbate from plasma, serum, urine, and fruit juices
- Assay 40 samples in duplicate
- Plate-based fluorometric measurement (ex 340 to 350 nm, em 420 to 430 nm)
Ascorbate (L-Ascorbic acid or Vitamin C) is a six-carbon lactone that is synthesized from glucose in the liver of most mammalian species, but not by humans who must obtain ascorbate through dietary consumption. In humans, ascorbate serves as an antioxidant and acts as an electron donor for various enzymes. Cayman’s Ascorbate Assay provides a reproducible, sensitive fluorescence-based tool for quantifying ascorbate from plasma, serum, urine, and fruit juices.