Order Entry
Denmark
ContactUsLinkComponent
Nitrotyrosine IP Kit
Nitrotyrosine IP Kit
Catalog # CAYM601220-1
Supplier:  Cayman Chemical
CAS Number:  
undefined
Nitrotyrosine IP Kit
Catalog # CAYM601220-1
Supplier:  Cayman Chemical
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Kit contains: Affinity sorbent, antibody, BSA, spin columns, and collection tubes
  • Brand name:
    CAYM
Specifications
About this item
For the capture and pulldown of nitrated proteins from cell and tissue lysates.
  • Determine change in tyrosine nitration in treated versus non-treated samples
  • Demonstrate target proteins are nitrated in vitro
  • Identify and characterise nitrated proteins by Western blot and proteomic analysis
Cayman’s Nitrotyrosine IP Kit allows for the capture and pulldown of nitrated proteins from cell lysates using a sorbent coupled with a nitrotyrosine monoclonal antibody. The nitrated proteins can be eluted from the resin, loaded to an SDS-PAGE gel, and probed using the included Nitrotyrosine Polyclonal Antibody or a user supplied target specific antibody. A positive control sample assures that the assay is working.