Order Entry
Denmark
ContactUsLinkComponent
QuickDetect™ Low Molecular Weight Heparin (LMWH) (Hu) ELISA Kit
QuickDetect™ Low Molecular Weight Heparin (LMWH) (Hu) ELISA Kit
Catalog # ABCAAB285268-96
Leverandør:  Abcam
CAS nummer:  
undefined
QuickDetect™ Low Molecular Weight Heparin (LMWH) (Hu) ELISA Kit
Catalog # ABCAAB285268-96
Leverandør:  Abcam
CAS nummer:  

Specifikationer

  • Pk:
    96 Tests
  • Assay Type:
    Sandwich
  • Description:
    QuickDetect™ low molecular weight heparin (LMWH) (Hu) ELISA kit
  • Tests per kit:
    100 assays

Specifikationer

Om denne vare

A rapid sandwich ELISA kit for quantifying of LMWH in human serum and other biological fluid.

  • Easy, convenient and time-saving
  • Performed within 90 minutes
  • Detection Range: 3,7 - 120 ng/mL

Low-molecular-weight heparin (LMWH) is a class of anticoagulant medications. They are used in the prevention blood clots and treatment of venous thromboembolism (deep vein thrombosis and pulmonary embolism) and in the treatment of myocardial infarction. It has less unpredictable side effect than naturally derived heparin.

BioVision's QuickDetectTM LMWH ELISA kit is a sandwich ELISA assay for measuring the quantity of human LMWH in serum, plasma and cell culture and other biological fluid. The entire process can be performed within 90 minutes with only a single incubation and a wash step respectively, resulting in fewer handling steps, fewer errors and more consistent results.

Cross Reactivity: No significant cross-reactivity or interference between this analyte and its analogues was observed.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.