Order Entry
Denmark
ContactUsLinkComponent
96-Well, Black Plates
96-Well, Black Plates
Catalog # ABCAAB286899-2
Supplier:  Abcam
undefined
96-Well, Black Plates
Catalog # ABCAAB286899-2
Supplier:  Abcam
Specifications
  • Description:
    96-well black plates
  • Pk:
    2
Specifications
About this item
Non-pyrogenic, polystyrene microplates (black) with standard solid flat bottom.
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Packaging dimension: 335×250×165 mm
  • The risk of sample carryover and cross-contamination is minimized
  • Suitable for fluorescence measurements
  • The polystyrene surface is more hydrophobic than the surface of a high binding microplate, and therefore tends to be more suitable for non-polar proteins and peptides
These are polystyrene microplates (solid black) with standard solid flat bottom, chimney well. The flat bottom allows precise optics and the rounded corners allow pipetting without leaving a residue.
Caution:
All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.