Order Entry
Denmark
ContactUsLinkComponent
Cleanroom Trousers, Movix Anti-static
Cleanroom Trousers, Movix Anti-static
Supplier:  ALSICO HIGHTECH
Certificates
undefined
Cleanroom Trousers, Movix Anti-static
Supplier:  ALSICO HIGHTECH
Specifications
  • Catalog No:
  • ALSO1033614-XL
  • ALSO1033614-L
  • ALSO1033614-3XL
  • ALSO1033614-XXL
  • ALSO1033614-XS
  • ALSO1033614-M
  • ALSO1033614-S
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Risk area:
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Fabric:
  • Movix Antistatic (96% polyester, 4% carbon)
  • Movix Antistatic (96% polyester, 4% carbon)
  • Movix Antistatic (96% polyester, 4% carbon)
  • Movix Antistatic (96% polyester, 4% carbon)
  • Movix Antistatic (96% polyester, 4% carbon)
  • Movix Antistatic (96% polyester, 4% carbon)
  • Movix Antistatic (96% polyester, 4% carbon)
  • Weight:
  • 152 g/m²
  • 152 g/m²
  • 152 g/m²
  • 152 g/m²
  • 152 g/m²
  • 152 g/m²
  • 152 g/m²