
4-Fluoro-1,3-dioxolan-2-one 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:114435-02-8
:
Formula:C₃H₃FO₃
MDL Number:MFCD06247543
:106.05 g/mol
Boiling Pt:212 °C
Melting Pt:18…23 °C
Flash Pt:>102 °C(216 °F)
Storage Temperature:Ambient
EINECS:483-360-5
:GHS07
Related Information
Frequently Bought Together