Order Entry
Denmark
ContactUsLinkComponent
Integrated suspension hoods, Versaflo™ S-400 Series
Integrated suspension hoods, Versaflo™ S-400 Series
Catalog # MMMAS-433L-5
Supplier:  3M
undefined
Integrated suspension hoods, Versaflo™ S-400 Series
Catalog # MMMAS-433L-5
Supplier:  3M

Specifications

  • Colour:
    White
  • Size:
    Large
  • Pk:
    5

Specifications

About this item

3M™ Versaflo™ S-400 series integrated suspension hood is a head cover with integrated head suspension which provides head, face, neck, and shoulder coverage. The hood can be used with 3M™ powered air respirators and supplied air respirator systems in a wide variety of applications.

  • Protects face and eyes from liquid splash and low energy flying particles and dust (Grade F)
  • For use with certain 3M™ Powered Air Purifying and Supplied Air Respirator Systems
  • Features the 3M™ Quick Release Swivel (QRS) connection
  • General-purpose two-layer polypropylene spunbond/laminated film

Get head, eye, and face protection with the Versaflo™ S-Series hood when working in food and beverage or pharmaceutical manufacturing. The integrated head suspension offers head, face, neck, and shoulder coverage and the eyes and face are shielded by the cover from liquid splash and low energy flying particles and dust.

This cost-effective headtop has been manufactured using a two-layer laminated film material. The integrated suspension makes it suitable for applications that require frequent replacement of the entire headtop assembly or where there is less soiling of the outer fabric. The suspension assembly and air inlet components can be easily removed and separated for segregated waste streams. There's no assembly required so it can be used straight out of the box.

Certifications: Headcover is CE approved and rated to EN 14594 3A, EN 12941 TH3 and EN 166:2:F:3.