Order Entry
Denmark
ContactUsLinkComponent
Bood DNA HV kit, CE IVD, Mag-Bind®
Bood DNA HV kit, CE IVD, Mag-Bind®
Catalog # OMEGM3292-03CEIVD
Supplier:  OMEGA BIO-TEK
CAS Number:  
undefined
Bood DNA HV kit, CE IVD, Mag-Bind®
Catalog # OMEGM3292-03CEIVD
Supplier:  OMEGA BIO-TEK
CAS Number:  

Specifications

  • Pk:
    96
  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Sample type:
    Blood
  • Target molecule:
    DNA
  • Description:
    Mag-Bind® blood DNA HV kit, CE IVD
  • No. of reactions:
    4×24 preparations

Specifications

About this item

The Mag-Bind® Blood DNA HV Kit CE IVD is designed for rapid and reliable isolation of high-quality genomic DNA from up to 4 ml whole blood samples.

The Mag-Bind® Blood DNA HV Kit CE IVD is designed for rapid and reliable isolation of high-quality genomic DNA from up to 4 ml whole blood samples. Mag-Bind® Particles CH provide a quick magnetic response time reducing overall processing time. This system combines the reversible nucleic acid-binding properties of Mag-Bind® paramagnetic particles with the time-proven efficiency of Omega Bio-Tek’s DNA isolation system to provide a fast and convenient method to isolate DNA. Utilizing paramagnetic particles provides high-quality DNA that is suitable for direct use in most downstream applications, such as amplification and enzymatic reactions. The 4 ml whole blood protocol has been successfully automated on the Hamilton Microlab® STAR™.

Certifications: CE-IVD IVDR