Polished mirrors
About this item
Our exceptionally hygienic mirrors are built to handle the toughest of environments.
- Shatterproof and safe
- De-burred edges and rounded corners to prevent injury
- 1,2 mm thick stainless steel
- Choose with or without pre-drilled mounting holes
- Vandal resistant
- Material: 304-grade stainless steel
Not only exceptionally hygienic, but also durable and long-lasting, our steel mirrors are made from 304-grade, meaning they can handle even the toughest of environments without breaking or shattering.
They have de-burred edges and rounded corners to prevent injury, ensuring your workspace is as safe as possible. The mirrors are also 1,2 mm thick, ensuring ultimate sturdiness and durability.
Also vandal-resistant, affording you maximum security and hygiene, ideal for your safe and conscious workplace.
As with all our products, our steel mirrors are suited to rigorous hygiene and HACCP routines, meaning you can rely on us to deliver a hygienic, durable and easy to clean product ideal for your workspace.
12 Options Available Below
Product Details & Documents
Our exceptionally hygienic mirrors are built to handle the toughest of environments.
- Shatterproof and safe
- De-burred edges and rounded corners to prevent injury
- 1,2 mm thick stainless steel
- Choose with or without pre-drilled mounting holes
- Vandal resistant
- Material: 304-grade stainless steel
Not only exceptionally hygienic, but also durable and long-lasting, our steel mirrors are made from 304-grade, meaning they can handle even the toughest of environments without breaking or shattering.
They have de-burred edges and rounded corners to prevent injury, ensuring your workspace is as safe as possible. The mirrors are also 1,2 mm thick, ensuring ultimate sturdiness and durability.
Also vandal-resistant, affording you maximum security and hygiene, ideal for your safe and conscious workplace.
As with all our products, our steel mirrors are suited to rigorous hygiene and HACCP routines, meaning you can rely on us to deliver a hygienic, durable and easy to clean product ideal for your workspace.
Product Family Options
Product Information
- AvailabilityPrice
- TEKMCRMI-600X400FHPolished mirror, rectangle with fixing holes, 600×400 mm
- TEKMCRMI-400X900FHPolished mirror, rectangle with fixing holes, 900×400 mm
- TEKMCRMI-PMFH18X60Polished mirror, rectangle with fixing holes, 1800×600 mm
- TEKMCRMI-300X600Polished mirror, rectangle, 600×300 mm
- TEKMCRMI-600X1200Polished mirror, rectangle, 1200×600 mm
- TEKMCRMI-300X1200Polished mirror, rectangle, 1200×300 mm
- TEKMCRMI-300X1500Polished mirror, rectangle, 1500×300 mm
- TEKMCRMI-600X1800Polished mirror, rectangle, 1800×600 mm
- TEKMCRMI-R450FHPolished mirror, round with fixing holes, 450 mm round
- TEKMCRMI-300X300Polished mirror, square, 300×300 mm
- TEKMCRMI-600X600Polished mirror, square, 600×600 mm
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



