Boxes for SCAT Politainer
Supplier: S.C.A.T.
Jump to:
About this item
Cardboard boxes for SCAT Politainer for liquid waste, compatible with the SCAT system.
- Space-saving storage
- Easy and safe to use
- Less expensive than comparable canisters
- Environmentally friendly disposal
Made of chemically resistant plastic, for many applications
Ordering information: The Politainers have to be ordered separately
3 Options Available Below
Product Details & Documents
Cardboard boxes for SCAT Politainer for liquid waste, compatible with the SCAT system.
- Space-saving storage
- Easy and safe to use
- Less expensive than comparable canisters
- Environmentally friendly disposal
Made of chemically resistant plastic, for many applications
Ordering information: The Politainers have to be ordered separately
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
Product Family Options
Product Information
- W×D×HForAvailabilityPrice
- SCTV107334Box, UN-Y approved
- SCTV107335Box, UN-Y approved
- SCTV107336Box, UN-Y approved
Politainers
Product Information
- Catalog #ThreadCapacityW×D×HTypeDescriptionColourAvailabilityPrice
- GL 385 L178×178×178 mmUN-Y approval in combination with boxPolitainerNaturalLog in for availability1 * 1 Items87,80 kr
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



