Lecithin, granular, for food industry
:8002-43-5
:
:MFCD00147406
:12,05452
:Ambient
- Environmentally Preferable:
- Acetone-Insoluble Matter:97.0 %
- Acid Value:max.35
- Hexane-Insoluble Matter:max. 0.05 %
- Water:max. 1.5 %
Frequently Bought Together