Potassium iodate ≥99.0% FCC
Catalog # SPCMP1309-25GM
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (KIO₃; Dried Basis):99.0 - 101.0 %
- Chlorate:To Pass Test (Li
- Iodide:To Pass Test (Li
- Lead (Pb):max. 4 mg/kg
- Loss On Drying:max. 0.5 %
Specification Test Results
About this item
:
:214 g/mol
UN:1479
:GHS03
:GHS07
Storage Temperature:Ambient
CAS Number:7758-05-6
Formula:KlO₃
MDL Number:MFCD00011406
Melting Pt:560
ADR:5.1 II
Merck Index:13,07726