Order Entry
Denmark
Orders LinkContactUsLinkComponent
Water agar

Water agar

Water agar, 500 g
Catalog # HIMEM1366-500G
Supplier:   HIMEDIA
Water agar
Catalog # HIMEM1366-500G
Supplier:  HIMEDIA
CAS Number:  
Water agar

Water agar, 500 g

CAS Number:  
Price
935,00 kr
Log in for availability
1 * 500 g
Product Details & Documents

Water Agar is recommended for enumeration, cultivation and observation of sporulation of some fungi.

  • Appearance: White to cream homogeneous free flowing powder
  • Gelling: Firm, comparable with 2,0% agar gel
  • Colour and clarity of prepared medium: White coloured clear to slightly opalescent gel forms in petri plates

Water agar is often recommended for enumeration, cultivation and observation of sporulation of some fungi

Water agar is used for enumeration of fungi according to MPN method.

Direct plating is considered to be one of the most effective techniques for mycological examination of all foods.

Specifications

Specifications

  • Pk:
    500 g
  • Description:
    Water agar

Specifications