TCBS agar
TCBS agar, 500 g
Catalog # HIMEM189-500G
Supplier: HIMEDIA
Jump to:
Product Details & Documents
TCBS agar, granulated is recommended for the selective isolation and cultivation of V. cholerae and other enteropathogenic Vibrio's causing food poisoning.
Specifications
Specifications
- Pk:500 g
- Description:TCBS agar
- Cat. No.:HIMEM189-500G
- Storage temperature:20...30 °C