Capping Reagent 90/10 Tetrahydrofuran/N-Methylimidazole (v/v), BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Catalog # HONCBR651-4X2.5L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 L
- Seq:5114
- Environmentally Preferable:
- Pk Info:4x 2.5 L
Specifications
About this item
:
Boiling Pt:66 °C (main component)
Melting Pt:–108.5 °C (main component)
Density:0.888 g/cm³ (20 °C) (main component)
Flash Pt:–15 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
:GHS02
:GHS08
:GHS05
:GHS07