Order Entry
Denmark
ContactUsLinkComponent
Capping Reagent, BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Irritant
Corrosive
Health
Flammable
Capping Reagent, BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™
Certificates
Irritant
Corrosive
Health
Flammable
Capping Reagent, BioSyn™ for DNA/RNA synthesis, Burdick & Jackson™

Specification Test Results

  • Appearance:
    complying
  • Identity (IR):
    complying
  • Assay Of Acetonitrile:
    79.0 - 81.0 Vol.%
  • N-methylimidazole (GC):
    19.0 - 21.0 Vol.%
  • Triazine:
    Max. 20 ppm
  • Water (Karl Fischer):
    Max. 50 ppm

Specification Test Results

Frequently Bought Together