Quintozene Analytical standard, reference material, Fluka™
Quintozene, Analytical standard, Glass Bottle, 100 mg
Catalog # HONCR1976-100MG
Supplier: Honeywell Chemicals
Quintozene Analytical standard, reference material, Fluka™
Catalog # HONCR1976-100MG
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard
- Description:Quintozene
- Cat. No.:HONCR1976-100MG
- Formula:C₆Cl₅NO₂
- Description:Quintozene
- Physical form:Solid
- MDL No.:MFCD00007065
- Melting point:143 - 145 °C
- Synonym(s):1,2,3,4,5-Pentachloro-6-nitrobenzene; Pentachloronitrobenzene
- Molecular weight:295.33 g/mol
- CAS No.:82-68-8



