Fluazuron Analytical standard, reference material, Fluka™
Fluazuron, Analytical standard, Glass Bottle, 100 mg
HONCR1973-100MG
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard
- Description:Fluazuron
- Cat. No.:HONCR1973-100MG
- Formula:C₂₀H₁₀Cl₂F₅N₃O₃
- Description:Fluazuron
- Physical form:Crystalline
- MDL No.:MFCD00864505
- Melting point:212 °C
- Synonym(s):1-(4-Chloro-3-{[3-chloro-5-(trifluoromethyl)-2-pyridyl]oxy}phenyl)-3-(2,6-difluorobenzoyl)urea; N-[[[4-chloro-3-[[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy]phenyl]amino]carbonyl]-2,6-difluorobenzamide
- Molecular weight:506.21 g/mol
- CAS No.:86811-58-7
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



