Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Ethyl laurate Analytical standard, reference material, Fluka™

Ethyl laurate, Analytical standard, Glass Bottle, 1 g
  HONCR1917-1G
Ethyl laurate Analytical standard, reference material, Fluka™
  HONCR1917-1G
 :  
Ethyl laurate Analytical standard, reference material, Fluka™

Ethyl laurate, Analytical standard, Glass Bottle, 1 g

 :  
 
393,00 kr
   
1 * 1 g
 

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery
 

 

  • Pack type:
    Glass Bottle
  • Pk:
    1 g
  • Standards:
    Analytical standard
  • Description:
    Ethyl laurate
  • Cat. No.:
    HONCR1917-1G
  • Formula:
    C₁₄H₂₈O₂
  • Description:
    Ethyl laurate
  • Storage conditions:
    RT
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00015065
  • Flash point:
    131 °C closed cup
  • Melting point:
    -10 °C
  • Synonym(s):
    Ethyl dodecanoate
  • Molecular weight:
    228.37 g/mol
  • CAS No.:
    106-33-2