Chlorothalonil Analytical standard, reference material, Fluka™
Chlorothalonil, Analytical standard, Glass Bottle, 100 mg
Catalog # HONCR1799-100MG
Supplier: Honeywell Chemicals
Chlorothalonil Analytical standard, reference material, Fluka™
Catalog # HONCR1799-100MG
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard
- Description:Chlorothalonil
- UN No.:2811
- Cat. No.:HONCR1799-100MG
- Description:Chlorothalonil
- Storage conditions:RT
- Physical form:Crystalline
- MDL No.:MFCD00045594
- Melting point:251 °C
- Molecular weight:265.91 g/mol
- CAS No.:1897-45-6
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



