Order Entry
Denmark
Orders LinkContactUsLinkComponent
Naproxen Sodium, Reference Standard

Naproxen Sodium, Reference Standard

Naproxen Sodium, USP reference standard, Vial, 200 mg
Catalog # 1457403.
Supplier:   USP
Naproxen Sodium, Reference Standard
Catalog # 1457403.
Supplier:  USP
CAS Number:  
Naproxen Sodium, Reference Standard

Naproxen Sodium, USP reference standard, Vial, 200 mg

CAS Number:  
Price
3.140,00 kr
Log in for availability
1 * 200 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    200 mg
  • Standards:
    USP reference standard
  • Description:
    Naproxen Sodium
  • Cat. No.:
    1457403.
  • Formula:
    C₁₄H₁₃NaO₃
  • Description:
    Naproxen Sodium
  • CAS No.:
    26159-34-2

Specifications