Order Entry
Denmark
Orders LinkContactUsLinkComponent
Dantrolene Related Compounds, Reference Standard

Dantrolene Related Compounds, Reference Standard

Supplier:   USP
Dantrolene Related Compounds, Reference Standard
Supplier:  USP
Certificates
Dantrolene Related Compounds, Reference Standard

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1163162.
  • 1163173.
  • 1163184.
  • Description:
  • Dantrolene Related Compound A
  • Dantrolene Related Compound B
  • Dantrolene Related Compound C
  • Synonym(s):
  • 1,2-Bis{[5-(4-nitrophenyl)furan-2-yl]methylene}hydrazine
  • 5-(4-nitrophenyl)-2-furaldehyde-(2-carboxymethyl) semicarbazone
  • 5-(4-Nitrophenyl)furan-2-carbaldehyde
  • CAS No.:
  • 301359-05-7
  • 57268-33-4
  • 7147-77-5
  • Formula:
  • C₂₂H₁₄N₄O₆
  • C₁₄H₁₂N₄O₆
  • C₁₁H₇NO₄

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1163162.
    Dantrolene Related Compound A
    Dantrolene Related Compound A
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 10.130,00 kr
    10.130,00 kr
    1 * 50 mg
       10.130,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    1163162.
    Description
    Dantrolene Related Compound A
    Synonym(s)
    1,2-Bis{[5-(4-nitrophenyl)furan-2-yl]methylene}hydrazine
    CAS No.
    301359-05-7
    Formula
    C₂₂H₁₄N₄O₆
  • 1163173.
    Dantrolene Related Compound B
    Dantrolene Related Compound B
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 10.020,00 kr
    10.020,00 kr
    1 * 50 mg
       10.020,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    1163173.
    Description
    Dantrolene Related Compound B
    Synonym(s)
    5-(4-nitrophenyl)-2-furaldehyde-(2-carboxymethyl) semicarbazone
    CAS No.
    57268-33-4
    Formula
    C₁₄H₁₂N₄O₆
  • 1163184.
    Dantrolene Related Compound C
    Dantrolene Related Compound C
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 9.940,00 kr
    9.940,00 kr
    1 * 50 mg
       9.940,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    1163184.
    Description
    Dantrolene Related Compound C
    Synonym(s)
    5-(4-Nitrophenyl)furan-2-carbaldehyde
    CAS No.
    7147-77-5
    Formula
    C₁₁H₇NO₄
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.