

Sodium metabisulphite mixture of NaHSO3 and Na2S2O5 ACS, Fluka™
Catalog # 243973-100G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Identity (IR):complying
- Assay of total-SO₂:min. 58.5 %
- Water insoluble matter:max. 0.005 %
- Iron (Fe):max. 0.002 %
- Heavy metals (as Pb):max. 0.001 %
- Chloride (Cl):max. 0.02 %
Specification Test Results
About this item
:
:190.11 g/mol
Boiling Pt:Decomposes on heating
Melting Pt:> 150 °C (dec.)
Density:2.36 g/cm³ (20 °C)
:GHS07
:GHS05
Storage Temperature:Room Temperature
CAS Number:7681-57-4
Formula:Na₂S₂O₅
MDL Number:MFCD00167602
Boiling Pt: Dec. (on heating)