Order Entry
Denmark
Orders LinkContactUsLinkComponent
Trypsin, Reference Standards

Trypsin, Reference Standards

Supplier:   USP
Trypsin, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1700013.
  • 1699956.
  • Description:
  • Trypsin Recombinant Porcine
  • Trypsin Bovine
  • CAS No.:
  • 9002-07-7
  • 9002-07-7
  • UN No.:
  • 1789

Specifications

Product Family Options

Product Information

  • Standards
    Pk Info
    Pack type
    Pk
    Availability
    Price
  • 1700013.
    Trypsin Recombinant Porcine
    Trypsin Recombinant Porcine
    USP reference standard
    2 x 1 mL
    Vial
    2
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - 17.960,00 kr
    17.960,00 kr
    1 * 2 Items
       17.960,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    1700013.
    Description
    Trypsin Recombinant Porcine
    CAS No.
    9002-07-7
    UN No.
    1789
  • 1699956.
    Trypsin Bovine
    Trypsin Bovine
    USP reference standard
    Vial
    300 mg
    Log in for availability
    Log in for availability
    1 * 300 mg
    1 * 300 mg - 6.490,00 kr
    6.490,00 kr
    1 * 300 mg
       6.490,00 kr

    Specifications:
    More
    Specifications
    Cat. No.
    1699956.
    Description
    Trypsin Bovine
    CAS No.
    9002-07-7
    UN No.