Order Entry
Denmark
Orders LinkContactUsLinkComponent
Neomycin, Reference Standards
 :   USP
Neomycin, Reference Standards
 :  USP
Neomycin, Reference Standards
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
 : US Pharmacopea Reference Standard
  • Catalog No:
  • 1458009.
  • 1458019.
  • 1458031.
  • 1458020.
  • Description:
  • Neomycin Sulfate
  • Neomycin B
  • Neomycin Sulfate System Suitability Mixture
  • Neomycin A
  • CAS No.:
  • 1405-10-3
  • 28002-70-2
  • 3947-65-7 (free base)
Product Family Options
  • Description
    Standards
    Pack type
    Pk
  • Neomycin Sulfate
    USP reference standard
    200 mg
    1 * 200 mg
    1 * 200 mg - 2.920,00 kr
    2.920,00 kr
    1 * 200 mg
       2.920,00 kr
    Specifications
    Cat. No.
    1458009.
    Description
    Neomycin Sulfate
    CAS No.
    1405-10-3
  • Neomycin B
    USP reference standard
    Vial
    150 mg
    1 * 150 mg
    1 * 150 mg - 3.080,00 kr
    3.080,00 kr
    1 * 150 mg
       3.080,00 kr
    Specifications
    Cat. No.
    1458019.
    Description
    Neomycin B
    CAS No.
    28002-70-2
  • Neomycin Sulfate System Suitability Mixture
    USP reference standard
    Vial
    100 mg
    1 * 100 mg
    1 * 100 mg - 3.510,00 kr
    3.510,00 kr
    1 * 100 mg
       3.510,00 kr
    Specifications
    Cat. No.
    1458031.
    Description
    Neomycin Sulfate System Suitability Mixture
    CAS No.
  • Neomycin A
    USP reference standard
    Vial
    2,5 mg
    1 * 2,5 mg
    1 * 2,5 mg - 4.820,00 kr
    4.820,00 kr
    1 * 2,5 mg
       4.820,00 kr
    Specifications
    Cat. No.
    1458020.
    Description
    Neomycin A
    CAS No.
    3947-65-7 (free base)