Bumetanide, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
3 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1078303.
- 1078325.
- 1078336.
- Description:
- Bumetanide
- Bumetanide Related Compound A
- Bumetanide Related Compound B
- Synonym(s):
- 3-Amino-4-phenoxy-5-sulfamoylbenzoic Acid
- 3-Nitro-4-phenoxy-5-sulfamoylbenzoic Acid
- CAS No.:
- 28395-03-1
- 28328-54-3
- 28328-53-2
- Formula:
- C₁₇H₂₀N₂O₅S
- C₁₃H₁₂N₂O₅S
- C₁₃H₁₀N₂O₇S
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1078303.BumetanideSpecifications:
More
Specifications
Cat. No.1078303.DescriptionBumetanideSynonym(s)CAS No.28395-03-1FormulaC₁₇H₂₀N₂O₅S - 1078325.Bumetanide Related Compound ABumetanide Related Compound AUSP reference standardVial10 mgLog in for availability1 * 10 mg9.260,00 krSpecifications:
More
Specifications
Cat. No.1078325.DescriptionBumetanide Related Compound ASynonym(s)3-Amino-4-phenoxy-5-sulfamoylbenzoic AcidCAS No.28328-54-3FormulaC₁₃H₁₂N₂O₅S - 1078336.Bumetanide Related Compound BBumetanide Related Compound BUSP reference standardVial25 mgLog in for availability1 * 25 mg9.730,00 krSpecifications:
More
Specifications
Cat. No.1078336.DescriptionBumetanide Related Compound BSynonym(s)3-Nitro-4-phenoxy-5-sulfamoylbenzoic AcidCAS No.28328-53-2FormulaC₁₃H₁₀N₂O₇S



