Acebutolol, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
3 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1000612.
- 1000623.
- 1000699.
- Description:
- Acebutolol Related Compound A
- Acebutolol Related Compound B
- Acebutolol
- Synonym(s):
- N-(3-Acetyl-4-hydroxyphenyl)butyramide
- N-{3-Acetyl-4-[2-hydroxy-3-(isopropylamino)propoxy]phenyl}acetamide
- N-{3-Acetyl-4-[3-(ethylamino)-2-hydroxypropoxy]phenyl}butyramide
- CAS No.:
- 40188-45-2
- 22568-64-5
- 441019-91-6
Specifications
Product Family Options
Product Information
- StandardsPkAvailabilityPrice
- 1000612.Acebutolol Related Compound AAcebutolol Related Compound AUSP reference standard20 mgLog in for availability1 * 20 mg5.580,00 krSpecifications:
More
Specifications
Cat. No.1000612.DescriptionAcebutolol Related Compound ASynonym(s)N-(3-Acetyl-4-hydroxyphenyl)butyramideCAS No.40188-45-2 - 1000623.Acebutolol Related Compound BAcebutolol Related Compound BUSP reference standard20 mgLog in for availability1 * 20 mg6.850,00 krSpecifications:
More
Specifications
Cat. No.1000623.DescriptionAcebutolol Related Compound BSynonym(s)N-{3-Acetyl-4-[2-hydroxy-3-(isopropylamino)propoxy]phenyl}acetamideCAS No.22568-64-5 - 1000699.AcebutololSpecifications:
More
Specifications
Cat. No.1000699.DescriptionAcebutololSynonym(s)N-{3-Acetyl-4-[3-(ethylamino)-2-hydroxypropoxy]phenyl}butyramideCAS No.441019-91-6
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



