PTFE tapes, ResTape
Supplier: Restek
Jump to:
About this item
ResTape PTFE tape is for use with threaded connections in a wide range of plumbing materials.
- Each roll is 12,7 mm × 6,6 m
Tapes have a maximum effective temperature of 260 °C.
3 Options Available Below
Product Details & Documents
ResTape PTFE tape is for use with threaded connections in a wide range of plumbing materials.
- Each roll is 12,7 mm × 6,6 m
Tapes have a maximum effective temperature of 260 °C.
Product Family Options
Product Information
- ColourAvailabilityPrice
- REST22485ResTape, compatible with gaseous or liquid oxygen, and with many other gases and liquidsResTape, compatible with gaseous or liquid oxygen, and with many other gases and liquidsGreenLog in for availability1 * 1 Items97,00 kr
- REST22486ResTape, compatible with a broad range of gases and liquidsResTape, compatible with a broad range of gases and liquidsYellowLog in for availability1 * 1 Items97,00 kr
- REST22487ResTape, anti-galling, compatible with stainless steel and many other metals and polymersResTape, anti-galling, compatible with stainless steel and many other metals and polymersGreyLog in for availability1 * 1 Items135,00 kr