
Sulpho-EMCS (N-(ε-maleimidocaproyloxy) sulphosuccinimide ester)
Catalog # BC17
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
Specification Test Results
- Spacer Arm (Å):9.4
- Reactive Toward:Primary Amine + Sulfhydryl
- Membrane Permeable:No
- Water Soluble:Yes
- Cleavable/ Reversible:No
Specification Test Results
About this item
:
:408.32 g/mol
:GHS07
Formula:C₁₄H₁₃O₉SN₂Na
MDL Number:MFCD09953186