Order Entry
Denmark
ContactUsLinkComponent
Desiccator cabinet, SICCO Star Thermo
Desiccator cabinet, SICCO Star Thermo
Catalog # BOHLV1830-07
Supplier:  Bohlender
undefined
Desiccator cabinet, SICCO Star Thermo
Catalog # BOHLV1830-07
Supplier:  Bohlender

Specifications

  • W×D×H ext.:
    310×420×630 mm
  • Description:
    Star-Thermo desiccator for drying and storage (55 L)
  • Environmentally Preferable:
  • Pk:
    1
  • Shelves:
    4
  • W×D×H int.:
    260×330×480 mm

Specifications

About this item

Aluminium frame with 2-pane insulation (inside glass, outside acrylic glass), including four shelves made of PC, usable with up to max. 26 shelves. Integrated heating (850 W) with temperature sensor and air circulation. Electrical power connection 230 V/50 Hz is required.

  • Reinforced aluminium frame with acrylic panels
  • Door with magnetic catch and circular rubber seal
  • Easy to read electronic hygrometer
  • Variable adjustable shelves
  • Extractable shelves
  • Non-slip rubber feet
  • UVC 1

SICCO Desiccators are designed for storing or drying humidity sensitive products using silica gel. The tight fitting door protects the contents from contamination from the atmosphere. The controlled environment inside the desiccators is ideal for storing reference materials, retained samples, metallographic specimen, tobacco and DNA-samples.