Order Entry
Denmark
ContactUsLinkComponent
 
EMCS (6-Maleimidohexanoic acid N-hydroxysuccinimide ester), Pierce™
  PIER22308
 :  
undefined
EMCS (6-Maleimidohexanoic acid N-hydroxysuccinimide ester), Pierce™
  PIER22308
 :  

 

  • Pk:
    50 mg
  • Seq:
    2370

 

 

  • Product Line:
    Pierce™
  • Labeling Method:
    Chemical Labeling
  • Crosslinker Type:
    Heterobifunctional
  • Reactive Moiety:
    Maleimide, NHS Ester
  • Spacer:
    Short
  • Form:
    Powder
  • Quantity:
    50 mg
  • Type:
    Crosslinker
  • Solubility:
    DMF, DMSO

 

 

 :
 :308.29 g/mol
 :Refrigerator
 :55750-63-5
 :C₁₄H₁₆N₂O₆
 :MFCD00043043