Order Entry
Denmark
ContactUsLinkComponent
Maltodextrin 12
Maltodextrin 12
Certificates
Maltodextrin 12
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White powder or granular powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤ 7.0%
  • pH:
    4.0 to 6.0 (1%, H2O)
  • Dextrose equivalent:
    9.0 to 15.0

Specification Test Results

Related Information

Frequently Bought Together