Coelenterazine ≥90% (by TLC), Calbiochem®, Sigma-Aldrich®
Catalog # 233900-250
Leverandør: Merck Millipore (Calbiochem)
CAS nummer:
Specifikationer
- Pack type:Plastic ampoule
- Pk:250 µG
- Seq:1370
Specifikationer
Om denne vare
:
:423.472 g/mol
:M-Clarity™ quality level = MQ100
Opbevaringstemperatur:Freezer
CAS nummer:55779-48-1
Formel:C₂₆H₂₁N₃O₃
MDL nummer:MFCD00467176