Sodium borohydride ≥97%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:16940-66-2
:
Formula:NaBH₄
MDL Number:MFCD00003518
:37.83 g/mol
Storage Temperature:Ambient
EINECS:241-004-4
UN:1426
ADR:4.3,I
:GHS02
:GHS06
:GHS05
:GHS08
Specification Test Results
- Assay :min. 97%
- Identity:passes test
- Appearance:white hygroscopic crystals
Specification Test Results
Related Information
Frequently Bought Together