Order Entry
Denmark
ContactUsLinkComponent
IMAP kinase, phosphatase, and phosphodiesterase assays, molecular devices
IMAP kinase, phosphatase, and phosphodiesterase assays, molecular devices
Catalog # MLDVR8160
Supplier:  Molecular Devices
CAS Number:  
IMAP kinase, phosphatase, and phosphodiesterase assays, molecular devices
Catalog # MLDVR8160
Supplier:  Molecular Devices
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    IMAP TR-FRET screening express kit with progressive binding system
  • Application:
    TR-FRET Screening

Specifications

About this item

IMAP® technology provides a homogeneous assay applicable to a wide variety of kinases without regard for substrate peptide sequence.

  • Complete assay system
  • Robust fluorescence signal
  • Homogeneous
  • Available in both FP and TR-FRET detection modes

IMAP provides a complete assay system for screening kinases, phosphatases, and phosphodiesterases

The assay is a simple "mix-and-read" procedure that allows accurate determination of kinase, phosphatase, and phosphodiesterase (PDE) activity.

IMAP Binding Reagent and Binding Buffers for 8000 data points in a standard 384 well format.