Order Entry
Denmark
ContactUsLinkComponent
Rat IgG2b Isotype Control [clone: LTF-2] (PE (Phycoerythrin)-Cy7®)
Rat IgG2b Isotype Control [clone: LTF-2] (PE (Phycoerythrin)-Cy7®)
Katalog # 60-4031-U100
Supplier:  Tonbo Biosciences
undefined
Rat IgG2b Isotype Control [clone: LTF-2] (PE (Phycoerythrin)-Cy7®)
Katalog # 60-4031-U100
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Control
  • Clone:
    LTF-2
  • Conjugation:
    PE (Phycoerythrin)-Cy7®
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2b
  • Storage temperature:
    Store Undiluted at 2...8 °C; Protect from Light; Do Not Freeze.
  • Tested applications:
    Flow Cytometry
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

The LTF-2 immunoglobulin is useful as an isotype-matched control. The LTF-2 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2b antibodies.