Order Entry
Denmark
ContactUsLinkComponent
 
Mouse IgG2a Isotype Control [clone: C1.18.4] (PE (Phycoerythrin)-Cy7®)
Catalog # 60-4724-U100
Supplier:  Tonbo Biosciences
undefined
Mouse IgG2a Isotype Control [clone: C1.18.4] (PE (Phycoerythrin)-Cy7®)
Catalog # 60-4724-U100
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Control
  • Clone:
    C1.18.4
  • Conjugation:
    PE (Phycoerythrin)-Cy7®
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Storage temperature:
    Store Undiluted at 2...8 °C; Protect from Light; Do Not Freeze.
  • Tested applications:
    Flow Cytometry
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

The C1.18.4 immunoglobulin is useful as an isotype-matched control. The C.1.18.4 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2a antibodies.

Type: Control
Antigen: C1.18.4
Clonality:
Clone: C1.18.4
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: 0