Order Entry
Denmark
ContactUsLinkComponent
CPC® DrumQuik® PUR Complete Drum Dispensing Systems
CPC® DrumQuik® PUR Complete Drum Dispensing Systems
Certificates
undefined
CPC® DrumQuik® PUR Complete Drum Dispensing Systems
Certificates
Specifications
  • Max. operating pressure:
  • 45 psi uncoupled, 22 psi coupled
  • 45 psi uncoupled, 22 psi coupled
  • 45 psi uncoupled, 22 psi coupled
  • 45 psi uncoupled, 22 psi coupled
  • Max. temperature:
  • 49 °C (120 °F)
  • 49 °C (120 °F)
  • 49 °C (120 °F)
  • 49 °C (120 °F)
  • Min. temperature:
  • 0 °C (32 °F)
  • 0 °C (32 °F)
  • 29 °C (–20 °F)
  • –18 °C (0 °F)
  • Seal material:
  • FKM
  • EPDM
  • FKM
  • Sterilisation:
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Autoclave or ethylene oxide (ETO)
  • Valve spring material:
  • PEEK
  • PEEK
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year