Ordreindtastning
Denmark
ContactUsLinkComponent
Parker Hannifin PFA Adapter Fittings, Compression to Male Threaded, Straight
Parker Hannifin Fitting, PFA, Straight, Compression to Threaded Adapter, 1/2" OD x 1/2" NPT(M)
Katalog # MFLX31320-13
Leverandør:  Avantor Fluid Handling
CE certificate
undefined
Parker Hannifin Fitting, PFA, Straight, Compression to Threaded Adapter, 1/2" OD x 1/2" NPT(M)
Katalog # MFLX31320-13
Leverandør:  Avantor Fluid Handling
CE certificate
Specifikationer
  • Tilslutningstype:
    Compression to Thread
  • Flow pattern:
    Straight
  • Thread size NPT(M) (port 2):
    ½"
  • Beskrivelse:
    Fitting, PFA, Straight, Compression to Threaded Adapter, ½" OD x ½" NPT(M)
  • Environmentally Preferable:
  • Sterilisering:
    Autoclave or ethylene oxide (ETO)
  • Garanti:
    18 months
  • Pakn:
    1
  • Fitting material:
    PFA
Specifikationer
Om denne vare
Superior chemical resistance with durable PFA.
  • Excellent choice to connect rigid tubing systems
  • Leak tight seal allows for less operation downtime
  • Ideal for general-purpose and single-use applications
These Parker Hannifin PFA compression to male thread adapter fittings are ideal for use in corrosive environments and chemical applications. Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
  • Tilslutningstype:
    Compression to Thread
  • Flow pattern:
    Straight
  • Thread size NPT(M) (port 2):
    ½"
  • Beskrivelse:
    Fitting, PFA, Straight, Compression to Threaded Adapter, ½" OD x ½" NPT(M)
  • Environmentally Preferable:
  • Sterilisering:
    Autoclave or ethylene oxide (ETO)
  • Garanti:
    18 months
  • Pakn:
    1
  • Fitting material:
    PFA