Order Entry
Denmark
ContactUsLinkComponent
 
Serum-free/low-protein medium, 1X with L-glutamine and 1 g/L BSA, Corning®
Catalog # MDTC40-101-CV
Supplier:  Corning
CAS Number:  
undefined
Serum-free/low-protein medium, 1X with L-glutamine and 1 g/L BSA, Corning®
Catalog # MDTC40-101-CV
Supplier:  Corning
CAS Number:  

Specifications

  • Pk:
    6
  • Description:
    Corning® cellgro® COMPLETE Serum-free/low-protein medium, 1X with L-Glutamine and 1 g/L BSA, media format - liquid
  • Size:
    500 mL
  • Storage temperature:
    Refrigerated
  • Shipping temperature:
    Ambient

Specifications

About this item

Corning® cellgro® COMPLETE formulation is based on a proprietary combination of 1:1 DMEM/F12, RPMI and McCoy's 5A medium. This medium contains a mixture of selected trace elements and high molecular weight carbohydrates, extra vitamins, animal origin-free protein, and a small amount of high quality bovine serum albumin (1,0 g/L). The Corning cellgro COMPLETE supports superior growth and viability over long-term passages in hybridomas, along with suspension and adherent cell cultures.