To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Power:120 VAC; 60 Hz
- Description:Explosion-proof electric motor, 9000 rpm, 120 VAC, 60 Hz
- Environmentally Preferable:
- CE regulatory approval:Yes
- UL US regulatory approval:Yes
- Warranty:3 Months
- Pk:1
- Max. motor RPM:9000 rpm
Configurable to your requirements.
- High-viscosity tube set handles fluids with viscosities to 50,000 cp!
Explosion-proof electric motors are rated for Class 1, Groups C and D hazardous environments and for pumping flammable liquids. Manual overload reset feature eliminates the danger of automatic restarting.
- Power:120 VAC; 60 Hz
- Description:Explosion-proof electric motor, 9000 rpm, 120 VAC, 60 Hz
- Environmentally Preferable:
- CE regulatory approval:Yes
- UL US regulatory approval:Yes
- Warranty:3 Months
- Pk:1
- Max. motor RPM:9000 rpm