Order Entry
Denmark
ContactUsLinkComponent
Penicillin : Streptomycin solution (20,000 IU Pen/mL, 20,000 µg Strep/mL), sterile
Health
Irritant
Penicillin : Streptomycin solution (20,000 IU Pen/mL, 20,000 µg Strep/mL), sterile
Catalog # 091670454.
Supplier:  MP Biomedicals
CAS Number:  
Health
Irritant
Penicillin : Streptomycin solution (20,000 IU Pen/mL, 20,000 µg Strep/mL), sterile
Catalog # 091670454.
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :GHS08
 :GHS07
Storage Temperature:Freezer
CAS Number:113-98-4 (Penicillin G potassium salt) - 5490-27-7 (Dihydrostreptomycin sulfate)
 :113-98-4