Legionella pneumophila (Serogroup 1) WDCM 00205 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Catalog # VT002057-10EA
Supplier: Merck
CAS Number:
Specifications
- Pk:10
- Description:Legionella pneumophila (serogroup 1) WDCM 00205 Vitroids™, >100’000 CFU mean value range
- M-Clarity quality level:MQ300
- Storage temperature:–20 °C
- Product type:Hydrosoluble discs
Specifications
About this item
Derived from CECT® strains.
- Origin Strain No. CECT® 8734
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.