Order Entry
Denmark
ContactUsLinkComponent
Enterococcus faecalis WDCM 00087 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Enterococcus faecalis WDCM 00087 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Catalog # VT000877-10EA
Supplier:  Merck
CAS Number:  
Enterococcus faecalis WDCM 00087 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Catalog # VT000877-10EA
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10
  • Description:
    Enterococcus faecalis WDCM 00087 Vitroids™, 80 - 130 CFU mean value range
  • M-Clarity quality level:
    MQ300
  • Storage temperature:
    –20 °C
  • Product type:
    Hydrosoluble discs

Specifications

About this item