Cronobacter sakazakii NCTC 11467 Low count LENTICULE® Discs 30-120 CFU Suitable for Microbiology, Certified Reference Material, Supelco®
Catalog # CRM11467L-10EA
Supplier: Merck
CAS Number:
Specifications
- Pk:10
- Description:Cronobacter sakazakii NCTC 11467 Low count LENTICULE® discs, 30 - 120 CFU
- M-Clarity quality level:MQ300
- Storage temperature:–20 °C
- Product type:Hydrosoluble discs
Specifications
About this item
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
Caution: LENTICULE® discs are manufactured by Sigma-Aldrich under licence from Public Health England.