Order Entry
Denmark
ContactUsLinkComponent
Clostridium perfringens NCTC 13170 Lenticule® Discs, Suitable for Microbiology, Certified Reference Material, Supelco®
Clostridium perfringens NCTC 13170 Lenticule® Discs, Suitable for Microbiology, Certified Reference Material, Supelco®
Catalog # CRM13170M-10EA
Supplier:  Merck
CAS Number:  
Clostridium perfringens NCTC 13170 Lenticule® Discs, Suitable for Microbiology, Certified Reference Material, Supelco®
Catalog # CRM13170M-10EA
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10
  • Description:
    Clostridium perfringens NCTC 13170 Lenticule® discs, 500-50000 CFU
  • M-Clarity quality level:
    MQ300
  • Storage temperature:
    –20 °C
  • Product type:
    Hydrosoluble discs

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
Caution: LENTICULE® discs are manufactured by Sigma-Aldrich under licence from Public Health England.